This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DDX46 SIRNA Assays ›
Gene Symbol:
Ddx46
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 46
Gene Aliases:
-
Chromosome Location:
Chr.17: 9475275 - 9519544 on Build Rnor_6.0
UniGene ID:
Rn.3436
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 193788

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ddx46
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139098.1 NP_620798.1 10 1364
XM_006253601.2 XP_006253663.1 10 1394
GenBank mRNA BC107590.1 - 10 1230 7461248
U25746.1 - 10 1364

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
helicase activity
ATP binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-9rq9x:80/100.66.78.177:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0