This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TCAM1 SIRNA Assays ›
Gene Symbol:
Tcam1
Gene Name:
testicular cell adhesion molecule 1
Gene Aliases:
Tcam1p
Chromosome Location:
Chr.10: 94465920 - 94478591 on Build Rnor_6.0
UniGene ID:
Rn.125075
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 194438

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tcam1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021673.1 NP_067705.1 7 1516
XM_006247642.2 XP_006247704.1 8 2270
GenBank mRNA BC166495.1 - 8 1710 7111123

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
single organismal cell-cell adhesion
integrin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline