siRNA Details
| Gene Symbol: Syt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030839.3 | NP_110466.2 | 2, 3 | 416 | ||
| XM_008762036.1 | XP_008760258.1 | 2, 3 | 1010 | |||
| GenBank mRNA | AF313453.1 | - | 2, 3 | 601 | ||
| AF375466.1 | - | 2, 3 | 416 | |||

Syt13
Genomic Map