siRNA Details
| Gene Symbol: Ralbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032067.1 | NP_114456.1 | 5 | 1390 | ||
| GenBank mRNA | BC089788.1 | - | 5 | 1444 | 7374343 | |
| U82623.1 | - | 5 | 1390 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Ralbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032067.1 | NP_114456.1 | 5 | 1390 | ||
| GenBank mRNA | BC089788.1 | - | 5 | 1444 | 7374343 | |
| U82623.1 | - | 5 | 1390 | |||
Ralbp1
Genomic Map