siRNA Details
| Gene Symbol: FBXO46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080469.1 | NP_001073938.1 | 2 | 2456 | ||
| GenBank mRNA | BC021978.2 | - | 1 | 1846 | 3957915 | |
| BC046245.1 | - | 1 | 1349 | |||
| Y10936.1 | - | 1 | 1002 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO46| Gene Symbol: FBXO46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080469.1 | NP_001073938.1 | 2 | 2456 | ||
| GenBank mRNA | BC021978.2 | - | 1 | 1846 | 3957915 | |
| BC046245.1 | - | 1 | 1349 | |||
| Y10936.1 | - | 1 | 1002 | |||