This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAPK8IP1 SIRNA Assays ›
Gene Symbol:
Mapk8ip1
Gene Name:
mitogen-activated protein kinase 8 interacting protein 1
Gene Aliases:
JIP1, Jip-1, Mapk8ip
Chromosome Location:
Chr.3: 81295023 - 81304181 on Build Rnor_6.0
UniGene ID:
Rn.44266
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 195747

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mapk8ip1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053777.1 NP_446229.1 2 302
GenBank mRNA AF108959.1 - 1 240
AF109772.1 - 1 149
AF109773.1 - 1 212
AF109774.1 - 2 302
DQ377223.1 - 1 112

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
signal transduction
JUN phosphorylation
positive regulation of signal transduction
vesicle-mediated transport
negative regulation of apoptotic process
negative regulation of JUN kinase activity
regulation of JNK cascade
negative regulation of JNK cascade
negative regulation of intrinsic apoptotic signaling pathway
MAP-kinase scaffold activity
protein binding
JUN kinase binding
kinesin binding
protein kinase binding
mitogen-activated protein kinase kinase binding
mitogen-activated protein kinase kinase kinase binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-d4kt8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline