siRNA Details
| Gene Symbol: Nat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037315.1 | NP_001032392.1 | 3 | 432 | ||
| NM_001037316.1 | NP_001032393.1 | 2 | 190 | |||
| NM_053853.2 | NP_446305.1 | 1 | 585 | |||
| XM_006253001.1 | XP_006253063.1 | 3 | 256 | |||
| XM_006253002.2 | XP_006253064.1 | 1 | 115 | |||
| XM_006253003.1 | XP_006253065.1 | 3 | 260 | |||
| XM_006253004.2 | XP_006253066.1 | 3 | 471 | |||
| GenBank mRNA | BC078765.1 | - | 2 | 191 | 7127165 | |
| CB580886.1 | - | 1 | 597 | |||
| U01343.1 | - | 3 | 432 | |||
| U01344.1 | - | 1 | 1083 | |||
| U17260.1 | - | 2 | 252 | |||

Nat1
Genomic Map