siRNA Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080480.2 | NP_536728.2 | 4 | 478 | ||
| GenBank mRNA | AF030558.1 | - | 4 | 478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pip4k2c| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080480.2 | NP_536728.2 | 4 | 478 | ||
| GenBank mRNA | AF030558.1 | - | 4 | 478 | ||