This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GALE SIRNA Assays ›
Gene Symbol:
Gale
Gene Name:
UDP-galactose-4-epimerase
Gene Aliases:
-
Chromosome Location:
Chr.5: 154310453 - 154314959 on Build Rnor_6.0
UniGene ID:
Rn.29908
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196011

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gale
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080783.2 NP_542961.2 3, 4 277
XM_006239134.2 XP_006239196.1 3, 4 1242
XM_008764197.1 XP_008762419.1 3, 4 508
GenBank mRNA BC097293.1 - 3, 4 277 7443707
X53949.1 - 2, 3 258

Back To Top

More Information


Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

galactose metabolic process
galactose catabolic process
UDP-N-acetylglucosamine 4-epimerase activity
UDP-glucose 4-epimerase activity
protein homodimerization activity
coenzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0