siRNA Details
| Gene Symbol: Gpm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178105.2 | NP_835206.1 | 6, 7 | 699 | ||
| XM_006253093.2 | XP_006253155.1 | 7, 8 | 1535 | |||
| GenBank mRNA | AB089242.1 | - | 6, 7 | 678 | ||
| BC088862.1 | - | 6, 7 | 699 | 7313006 | ||
| FQ211804.1 | - | 7, 8 | 1154 | |||

Gpm6a
Genomic Map