siRNA Details
| Gene Symbol: Ctsj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017121.1 | NP_058817.1 | 5 | 642 | ||
| XM_008771439.1 | XP_008769661.1 | 4 | 557 | |||
| GenBank mRNA | AF310623.2 | - | 5 | 642 | ||
| BC097263.1 | - | 5 | 625 | 7460052 | ||
| L14776.1 | - | 2 | 268 | |||

Ctsj
Genomic Map