siRNA Details
| Gene Symbol: Tsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019203.1 | NP_062076.1 | 4 | 293 | ||
| XM_006257142.2 | XP_006257204.1 | 5 | 373 | |||
| GenBank mRNA | X99797.1 | - | 4 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsx| Gene Symbol: Tsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019203.1 | NP_062076.1 | 4 | 293 | ||
| XM_006257142.2 | XP_006257204.1 | 5 | 373 | |||
| GenBank mRNA | X99797.1 | - | 4 | 293 | ||