This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C1QBP SIRNA Assays ›
Gene Symbol:
C1qbp
Gene Name:
complement component 1, q subcomponent binding protein
Gene Aliases:
Habp1, gC1qR
Chromosome Location:
Chr.10: 57613876 - 57618527 on Build Rnor_6.0
UniGene ID:
Rn.2765
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198852

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1qbp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019259.2 NP_062132.2 2 297
GenBank mRNA AJ001102.1 - 2 338
BC070510.1 - 2 297 7100505
FQ212622.1 - 2 322
FQ214537.1 - 2 347
FQ219796.1 - 2 320
FQ220621.1 - 2 340

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
mRNA processing
apoptotic process
immune response
complement activation, classical pathway
RNA splicing
phosphatidylinositol 3-kinase signaling
regulation of complement activation
negative regulation of interferon-gamma production
negative regulation of interleukin-12 production
negative regulation of MDA-5 signaling pathway
negative regulation of RIG-I signaling pathway
mature ribosome assembly
positive regulation of apoptotic process
innate immune response
positive regulation of cell adhesion
negative regulation of mRNA splicing, via spliceosome
negative regulation of defense response to virus
positive regulation of protein kinase B signaling
positive regulation of mitochondrial translation
positive regulation of neutrophil chemotaxis
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of trophoblast cell migration
positive regulation of dendritic cell chemotaxis
complement component C1q binding
transcription corepressor activity
mRNA binding
protein kinase C binding
protein binding
hyaluronic acid binding
transcription factor binding
kininogen binding
adrenergic receptor binding
mitochondrial ribosome binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline