This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KCNN1 SIRNA Assays ›
Gene Symbol:
Kcnn1
Gene Name:
potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1
Gene Aliases:
KCa2.1
Chromosome Location:
Chr.16: 20336784 - 20349168 on Build Rnor_6.0
UniGene ID:
Rn.44422
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198973

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019313.1 NP_062186.1 5, 6 1159
XM_006252933.2 XP_006252995.1 5, 6 1192
GenBank mRNA AF000973.1 - 5, 6 1159
DQ137426.1 - 1, 2 114
U69885.1 - 5, 6 925

Back To Top

More Information


Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transmembrane transport
calmodulin binding
small conductance calcium-activated potassium channel activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline