siRNA Details
| Gene Symbol: Park2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020093.1 | NP_064478.1 | 3 | 258 | ||
| GenBank mRNA | AB039878.1 | - | 3 | 258 | ||
| AF168004.1 | - | 3 | 278 | |||
| AF210434.1 | - | 3 | 250 | |||
| AF257234.1 | - | 2 | 188 | |||
| AF343575.1 | - | 1 | 24 | |||
| AF381277.1 | - | 3 | 260 | |||
| AF381278.1 | - | 2 | 188 | |||
| AF381279.1 | - | 2 | 188 | |||
| AF381280.1 | - | 3 | 344 | |||
| AF381281.1 | - | 2 | 188 | |||
| GU322017.1 | - | 4 | 357 | |||
| GU322019.1 | - | 3 | 201 | |||
| GU322363.1 | - | 3 | 201 | |||
| GU322364.1 | - | 3 | 201 | |||
| GU345835.1 | - | 3 | 201 | |||
| GU345836.1 | - | 3 | 201 | |||
| KC774169.1 | - | 2 | 44 | |||
| KC774170.1 | - | 2 | 44 | |||
| KC774172.1 | - | 3 | 198 | |||
| KC774173.1 | - | 2 | 44 | |||
| KC774175.1 | - | 2 | 44 | |||
| KC774177.1 | - | 2 | 44 | |||

Park2
Genomic Map