siRNA Details
| Gene Symbol: Pde1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022710.1 | NP_073201.1 | 6 | 624 | ||
| XM_006242386.2 | XP_006242448.1 | 5 | 1196 | |||
| GenBank mRNA | AF327906.1 | - | 6 | 617 | ||
| FQ212460.1 | - | 6 | 680 | |||
| M94537.1 | - | 6 | 624 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001285890, NM_008800, L01695, BC058531, AK004772.

Pde1b
Genomic Map