siRNA Details
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012670.1 | NP_036802.1 | 4 | 382 | ||
| GenBank mRNA | D90345.1 | - | 4 | 382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcp1| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012670.1 | NP_036802.1 | 4 | 382 | ||
| GenBank mRNA | D90345.1 | - | 4 | 382 | ||