This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GFRA2 SIRNA Assays ›
Gene Symbol:
Gfra2
Gene Name:
GDNF family receptor alpha 2
Gene Aliases:
Retl2
Chromosome Location:
Chr.15: 52557607 - 52648973 on Build Rnor_6.0
UniGene ID:
Rn.90137
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 200352

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gfra2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012750.1 NP_036882.1 3 488
XM_006252238.2 XP_006252300.1 2 1257
GenBank mRNA AF003825.1 - 1038
AF005226.1 - 3 368
AY059645.1 - 2 38
U97143.1 - 3 488

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

transmembrane receptor protein tyrosine kinase signaling pathway
nervous system development
negative regulation of protein autophosphorylation
glial cell-derived neurotrophic factor receptor signaling pathway
glial cell-derived neurotrophic factor receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline