siRNA Details
| Gene Symbol: Abcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022281.2 | NP_071617.2 | 31 | 4778 | ||
| GenBank mRNA | AF487549.1 | - | 31 | 4778 | ||
| AJ277881.1 | - | 15 | 2609 | |||
| AY174892.1 | - | 31 | 4751 | |||
| AY174893.1 | - | 30 | 4651 | |||
| X96394.1 | - | 3 | 466 | |||
| Gene Symbol: LOC688708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767477.1 | XP_008765699.1 | 14 | 3323 | ||

Abcc1
Genomic Map