siRNA Details
| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031514.1 | NP_113702.1 | 3 | 312 | ||
| GenBank mRNA | U13396.1 | - | 3 | 312 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jak2| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031514.1 | NP_113702.1 | 3 | 312 | ||
| GenBank mRNA | U13396.1 | - | 3 | 312 | ||