This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PCSK9 SIRNA Assays ›
Gene Symbol:
Pcsk9
Gene Name:
proprotein convertase subtilisin/kexin type 9
Gene Aliases:
NARC-1, Narc1, PC9
Chromosome Location:
Chr.5: 126031368 - 126053726 on Build Rnor_6.0
UniGene ID:
Rn.19195
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 201747

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pcsk9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199253.2 NP_954862.2 1 342
GenBank mRNA AY847775.1 - 1 342
BC133063.1 - 1 273 8371686

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

kidney development
liver development
negative regulation of receptor recycling
positive regulation of receptor internalization
proteolysis
triglyceride metabolic process
phospholipid metabolic process
apoptotic process
lysosomal transport
cholesterol metabolic process
cellular response to starvation
regulation of receptor activity
negative regulation of low-density lipoprotein particle clearance
protein processing
protein autoprocessing
neurogenesis
neuron differentiation
low-density lipoprotein receptor particle metabolic process
low-density lipoprotein particle receptor catabolic process
regulation of low-density lipoprotein particle receptor catabolic process
cellular response to insulin stimulus
lipoprotein metabolic process
cholesterol homeostasis
regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
serine-type endopeptidase activity
sodium channel inhibitor activity
low-density lipoprotein particle binding
apolipoprotein binding
very-low-density lipoprotein particle binding
apolipoprotein receptor binding
protein self-association
poly(A) RNA binding
low-density lipoprotein particle receptor binding
very-low-density lipoprotein particle receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline