siRNA Details
| Gene Symbol: Slc4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270794.1 | NP_001257723.1 | 2 | 212 | ||
| NM_001270795.1 | NP_001257724.1 | 2 | 212 | |||
| NM_199497.2 | NP_955791.1 | 2 | 212 | |||
| GenBank mRNA | AY489024.1 | - | 2 | 212 | ||
| AY489025.1 | - | 2 | 212 | |||
| AY489026.1 | - | 2 | 212 | |||

Slc4a8
Genomic Map