This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADAM23 SIRNA Assays ›
Gene Symbol:
ADAM23
Gene Name:
ADAM metallopeptidase domain 23
Gene Aliases:
MDC-3, MDC3
Chromosome Location:
Chr.2: 206443544 - 206621130 on Build GRCh38
UniGene ID:
Hs.370287
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 202065

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ADAM23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003812.3 NP_003803.1 12 1468
XM_005246932.3 XP_005246989.1 12 1568
XM_011512086.2 XP_011510388.1 12 1568
GenBank mRNA AB009672.1 - 12 1468
AJ005580.1 - 12 1297
AK129906.1 - 8 632
AK296844.1 - 12 1177
BC132763.1 - 12 1448 40146785
BC132765.1 - 12 1448 40146787

Back To Top

More Information


Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cell adhesion
central nervous system development
metalloendopeptidase activity
integrin binding
protein binding
metallopeptidase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline