This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TAS2R8 SIRNA Assays ›
Gene Symbol:
TAS2R8
Gene Name:
taste 2 receptor member 8
Gene Aliases:
T2R8, TRB5
Chromosome Location:
Chr.12: 10806051 - 10806980 on Build GRCh38
UniGene ID:
Hs.533755
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 202287

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TAS2R8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023918.1 NP_076407.1 1 890
GenBank mRNA BC069084.1 - 1 890
BC069551.1 - 1 890 7262241
BC096701.1 - 1 890 40015728
BC096735.1 - 1 890 40015730
BC098167.1 - 1 890 40015729
BC098353.1 - 1 890 40015727

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
taste receptor activity
bitter taste receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-sqn2v:80/100.66.78.101:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0