siRNA Details
| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005304.3 | NP_005295.1 | 2 | 1237 | ||
| XM_011526858.2 | XP_011525160.1 | 2 | 1593 | |||
| GenBank mRNA | AK290710.1 | - | 2 | 1237 | ||
| BC035657.1 | - | 2 | 1223 | 5588752 | ||

FFAR3
Genomic Map