siRNA Details
| Gene Symbol: UBIAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013319.2 | NP_037451.1 | 1 | 698 | ||
| XM_006710590.3 | XP_006710653.1 | 1 | 698 | |||
| XM_011541304.2 | XP_011539606.1 | 1 | 698 | |||
| GenBank mRNA | AF117064.1 | - | 1 | 696 | ||
| AK074890.1 | - | 1 | 690 | |||
| AK222808.1 | - | 1 | 698 | |||
| BC004468.2 | - | 1 | 390 | 3936638 | ||
| BT006832.1 | - | 1 | 364 | |||

UBIAD1
Genomic Map