siRNA Details
| Gene Symbol: LINC00652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF161557.1 | - | 3 | 454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00652| Gene Symbol: LINC00652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF161557.1 | - | 3 | 454 | ||