This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other UNC50 SIRNA Assays ›
Gene Symbol:
UNC50
Gene Name:
unc-50 inner nuclear membrane RNA binding protein
Gene Aliases:
GMH1, HSD23, UNCL, URP
Chromosome Location:
Chr.2: 98608569 - 98618515 on Build GRCh38
UniGene ID:
Hs.13370
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 20778

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UNC50
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014044.5 NP_054763.2 6 1092
XM_005263913.4 XP_005263970.1 6 1188
XM_006712403.3 XP_006712466.1 6 1210
XM_006712404.2 XP_006712467.1 6 1124
GenBank mRNA AF077038.1 - 6 1044
AK222599.1 - 6 1058
AL080115.1 - 6 1096
AY017215.1 - 6 1060
BC017019.1 - 6 1225 3888349
BC025992.2 - 5 2120 4830706
BC039078.1 - 5 2096 5166015
BC091481.1 - 6 1082 30337350

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
cell surface receptor signaling pathway
protein transport
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5wqn7:80/100.66.79.161:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline