This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OR2AG1 SIRNA Assays ›
Gene Symbol:
OR2AG1 OR2AG2
Gene Name:
olfactory receptor family 2 subfamily AG member 1 (gene/pseudogene)
olfactory receptor family 2 subfamily AG member 2
Gene Aliases:
OR11-79, OR2AG3
OR11-76, OR2AG2P
Chromosome Location:
Chr.11: 6785017 - 6786014 on Build GRCh38
Chromosome Location:
Chr.11: 6768007 - 6768957 on Build GRCh38
UniGene ID:
Hs.553608
Hs.534614
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 213095

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR2AG1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004489.2 NP_001004489.1 1 186
GenBank mRNA BC151143.1 - 1 186 9020805
Gene Symbol: OR2AG2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004490.1 NP_001004490.1 1 165

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4m7g7:80/100.66.75.155:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline