siRNA Details
| Gene Symbol: OR8G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291438.1 | NP_001278367.1 | 1 | 108 | ||
| GenBank mRNA | BC136638.1 | - | 1 | 108 | 9020628 | |
| BC136639.1 | - | 1 | 108 | 9020629 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR8G2| Gene Symbol: OR8G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291438.1 | NP_001278367.1 | 1 | 108 | ||
| GenBank mRNA | BC136638.1 | - | 1 | 108 | 9020628 | |
| BC136639.1 | - | 1 | 108 | 9020629 | ||