This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other UBE2M SIRNA Assays ›
Gene Symbol:
UBE2M UBE2MP1
Gene Name:
ubiquitin conjugating enzyme E2 M
ubiquitin conjugating enzyme E2 M pseudogene 1
Gene Aliases:
UBC-RS2, UBC12, hUbc12
-
Chromosome Location:
Chr.19: 58555712 - 58558976 on Build GRCh38
Chromosome Location:
Chr.16: 35169431 - 35170391 on Build GRCh38
UniGene ID:
Hs.406068
Hs.646441
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 213508

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UBE2M
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003969.3 NP_003960.1 2 766
GenBank mRNA AB012191.1 - 2 171
AF075599.1 - 2 174
BC058924.1 - 2 280 6162220
BT006754.1 - 2 171
BU508830.1 - 2 710
Gene Symbol: UBE2MP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002837.1 1 320
GenBank mRNA AW376368.1 - 1 122

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
transforming growth factor beta receptor signaling pathway
protein ubiquitination
NIK/NF-kappaB signaling
positive regulation of neuron apoptotic process
protein neddylation
ubiquitin-protein transferase activity
protein binding
ATP binding
ribosomal S6-glutamic acid ligase activity
NEDD8 transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline