This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SKP2 SIRNA Assays ›
Gene Symbol:
SKP2
Gene Name:
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
Gene Aliases:
FBL1, FBXL1, FLB1, p45
Chromosome Location:
Chr.5: 36151989 - 36192792 on Build GRCh38
UniGene ID:
Hs.23348
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 213582

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SKP2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005983.3 NP_005974.2 2, 3 498
NM_032637.3 NP_116026.1 2, 3 498
XM_011514081.2 XP_011512383.1 2, 3 552
XM_011514082.2 XP_011512384.1 2, 3 552
GenBank mRNA AB050979.1 - 1, 2 278
AB050980.1 - 1, 2 278
AB050981.1 - 1, 2 278
AK291255.1 - 2, 3 430
AK295152.1 - 2, 3 372
AY029177.1 - 2, 3 330
BC001441.2 - 2, 3 417 3138709
BC007441.2 - 2, 3 421 2962938
DA223617.1 - 498
U33761.1 - 2, 3 461

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein polyubiquitination
cell proliferation
anaphase-promoting complex-dependent catabolic process
positive regulation of intracellular estrogen receptor signaling pathway
regulation of apoptotic process
positive regulation of smooth muscle cell proliferation
regulation of cell cycle
cellular response to cell-matrix adhesion
positive regulation of protein polyubiquitination
ubiquitin-protein transferase activity
protein binding
identical protein binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-m6gg6:80/100.66.76.251:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0