This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BIRC6 SIRNA Assays ›
Gene Symbol:
BIRC6
Gene Name:
baculoviral IAP repeat containing 6
Gene Aliases:
APOLLON, BRUCE
Chromosome Location:
Chr.2: 32356907 - 32618899 on Build GRCh38
UniGene ID:
Hs.150107
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 213978

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BIRC6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016252.3 NP_057336.3 11 3122
XM_005264449.4 XP_005264506.2 11 3243
XM_005264450.4 XP_005264507.2 11 3243
XM_005264451.4 XP_005264508.2 11 3243
XM_005264452.4 XP_005264509.2 11 2988
XM_005264453.4 XP_005264510.2 11 2988
XM_005264454.4 XP_005264511.2 11 3243
XM_005264455.4 XP_005264512.2 11 3243
XM_006712054.3 XP_006712117.1 11 3243
XM_006712055.3 XP_006712118.1 11 3243
XM_006712056.3 XP_006712119.1 11 3243
XM_011533003.2 XP_011531305.1 11 2688
XM_011533005.2 XP_011531307.1 11 3243
XM_017004556.1 XP_016860045.1 11 3243
XM_017004557.1 XP_016860046.1 11 3243
XM_017004558.1 XP_016860047.1 11 3243
XM_017004559.1 XP_016860048.1 11 3243
XM_017004560.1 XP_016860049.1 11 3243
GenBank mRNA AF265555.1 - 11 2904

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
mitotic nuclear division
positive regulation of cell proliferation
negative regulation of endopeptidase activity
protein ubiquitination
regulation of cytokinesis
regulation of cell proliferation
negative regulation of apoptotic process
cell division
labyrinthine layer development
spongiotrophoblast layer development
negative regulation of extrinsic apoptotic signaling pathway
ubiquitin-protein transferase activity
cysteine-type endopeptidase inhibitor activity
protein binding
ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline