This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PBK SIRNA Assays ›
Gene Symbol:
PBK
Gene Name:
PDZ binding kinase
Gene Aliases:
CT84, HEL164, Nori-3, SPK, TOPK
Chromosome Location:
Chr.8: 27809621 - 27838095 on Build GRCh38
UniGene ID:
Hs.104741
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 214040

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PBK
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278945.1 NP_001265874.1 7 1050
NM_018492.3 NP_060962.2 7 1075
XM_006716368.2 XP_006716431.1 7 999
GenBank mRNA AB027249.1 - 7 801
AB027250.1 - 7 781
AF189722.1 - 7 765
AF237709.1 - 7 789
AK027291.1 - 7 780
AK301836.1 - 7 1010
BC015191.1 - 7 789 4082846
EU794685.1 - 7 812

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
mitotic nuclear division
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of stress-activated MAPK cascade
cellular response to UV
negative regulation of inflammatory response
protein serine/threonine kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0