This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PEG10 SIRNA Assays ›
Gene Symbol:
PEG10
Gene Name:
paternally expressed 10
Gene Aliases:
EDR, HB-1, MEF3L, Mar2, Mart2, RGAG3
Chromosome Location:
Chr.7: 94656325 - 94669695 on Build GRCh38
UniGene ID:
Hs.147492
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 214912

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PEG10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040152.1 NP_001035242.1 2 271
NM_001172437.2 NP_001165908.1 2 282
NM_001172438.2 NP_001165909.1 2 282
NM_001184961.1 NP_001171890.1 2 271
NM_001184962.1 NP_001171891.1 2 271
NM_015068.3 NP_055883.2 2 271
GenBank mRNA AB049150.1 - 2 50
AK299837.1 - 237
AL589326.1 - 2 253
BC050659.2 - 2 203 6197322
BP250746.1 - 2 271

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
cell differentiation
negative regulation of transforming growth factor beta receptor signaling pathway
DNA binding
protein binding
zinc ion binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline