This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OR10Q1 SIRNA Assays ›
Gene Symbol:
OR10Q1
Gene Name:
olfactory receptor family 10 subfamily Q member 1
Gene Aliases:
OR11-233
Chromosome Location:
Chr.11: 58227882 - 58228918 on Build GRCh38
UniGene ID:
Hs.553646
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 215343

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR10Q1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004471.2 NP_001004471.1 1 576
GenBank mRNA BC140731.1 - 1 576 9021599

Back To Top

More Information


Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0