This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF740 SIRNA Assays ›
Gene Symbol:
ZNF740
Gene Name:
zinc finger protein 740
Gene Aliases:
Zfp740
Chromosome Location:
Chr.12: 53180715 - 53190870 on Build GRCh38
UniGene ID:
Hs.524458
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 215391

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF740
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004304.3 NP_001004304.1 3, 4 561
XM_006719344.2 XP_006719407.1 3, 4 536
XM_011538194.2 XP_011536496.1 3, 4 761
XM_017019181.1 XP_016874670.1 3, 4 795
GenBank mRNA AK292744.1 - 3, 4 612
BC053557.1 - 3, 4 564 6162269

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleic acid binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-cdc4cf686-kglqn:80/100.66.78.176:80.
git-commit: 18f7f941bc72f70e8ff4e0be257f7363f2314623
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.0-Offline