siRNA Details
| Gene Symbol: KRTAP21-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181617.1 | NP_853648.1 | 1 | 240 | ||
| GenBank mRNA | AB096959.1 | - | 1 | 272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP21-2| Gene Symbol: KRTAP21-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181617.1 | NP_853648.1 | 1 | 240 | ||
| GenBank mRNA | AB096959.1 | - | 1 | 272 | ||