siRNA Details
| Gene Symbol: KRTAP22-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181620.1 | NP_853651.1 | 1 | 110 | ||
| GenBank mRNA | AB096953.1 | - | 1 | 144 | ||
| BC101682.1 | - | 1 | 136 | 8069188 | ||
| BC101686.1 | - | 1 | 136 | 8069192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP22-1| Gene Symbol: KRTAP22-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181620.1 | NP_853651.1 | 1 | 110 | ||
| GenBank mRNA | AB096953.1 | - | 1 | 144 | ||
| BC101682.1 | - | 1 | 136 | 8069188 | ||
| BC101686.1 | - | 1 | 136 | 8069192 | ||