This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF383 SIRNA Assays ›
Gene Symbol:
ZNF383
Gene Name:
zinc finger protein 383
Gene Aliases:
HSD17
Chromosome Location:
Chr.19: 37217243 - 37248740 on Build GRCh38
UniGene ID:
Hs.590975
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 215883

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF383
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152604.1 NP_689817.1 5 1163
XM_005258585.2 XP_005258642.1 6 1421
XM_005258587.2 XP_005258644.1 5 1298
XM_005258588.2 XP_005258645.1 6 1060
XM_011526586.2 XP_011524888.1 7 2166
XM_011526587.2 XP_011524889.1 6 2133
XM_011526588.2 XP_011524890.1 5 2315
XM_011526589.2 XP_011524891.1 5 996
XM_011526590.2 XP_011524892.1 4 961
XM_017026422.1 XP_016881911.1 7 1572
XM_017026423.1 XP_016881912.1 7 2166
XM_017026424.1 XP_016881913.1 5 1212
GenBank mRNA AK091760.1 - 5 1724
AK093182.1 - 5 1163
AK122892.1 - 8 1489
AY260141.1 - 4 958
AY438646.1 - 5 1163

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-sdrxx:80/100.66.78.100:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline