siRNA Details
| Gene Symbol: NRROS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198565.2 | NP_940967.1 | 3 | 1577 | ||
| GenBank mRNA | AY358322.1 | - | 3 | 1483 | ||
| BC044233.1 | - | 3 | 1488 | 5725567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NRROS| Gene Symbol: NRROS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198565.2 | NP_940967.1 | 3 | 1577 | ||
| GenBank mRNA | AY358322.1 | - | 3 | 1483 | ||
| BC044233.1 | - | 3 | 1488 | 5725567 | ||