This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM43B SIRNA Assays ›
Gene Symbol:
FAM43B
Gene Name:
family with sequence similarity 43 member B
Gene Aliases:
-
Chromosome Location:
Chr.1: 20552439 - 20555020 on Build GRCh38
UniGene ID:
Hs.429179
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 216109

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM43B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207334.2 NP_997217.1 1 2320
GenBank mRNA AK126900.1 - 1 2320
BC142610.1 - 1 1963 40147211
BC142686.1 - 1 2211 40148591

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nbfjs:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline