This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FASTKD3 SIRNA Assays ›
Gene Symbol:
FASTKD3
Gene Name:
FAST kinase domains 3
Gene Aliases:
-
Chromosome Location:
Chr.5: 7859159 - 7869037 on Build GRCh38
UniGene ID:
Hs.653162
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 216139

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FASTKD3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024091.3 NP_076996.2 4 1861
NR_036553.1 3 310
NR_073608.1 3 310
XM_006714498.1 XP_006714561.1 4 1772
GenBank mRNA AF161354.1 - 3 284
AK026927.1 - 4 1826
BC001295.1 - 3 1353
BC113563.1 - 3 1776 8322615
BC143517.1 - 3 1776
BC143518.1 - 4 1213
BG613096.1 - 3 300

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cellular respiration
protein kinase activity
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-gbfjn:80/100.66.78.155:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0