This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other JPH2 SIRNA Assays ›
Gene Symbol:
JPH2
Gene Name:
junctophilin 2
Gene Aliases:
CMH17, JP-2, JP2
Chromosome Location:
Chr.20: 44111697 - 44187578 on Build GRCh38
UniGene ID:
Hs.441737
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 216294

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: JPH2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020433.4 NP_065166.2 1 1057
NM_175913.3 NP_787109.2 1 1057
XM_006723833.3 XP_006723896.1 1 1049
GenBank mRNA AL132999.1 - 1 572
BC029957.1 - 1 1057

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

release of sequestered calcium ion into cytosol
regulation of cardiac muscle tissue development
calcium ion homeostasis
regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
calcium ion transport into cytosol
phosphatidylserine binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
phosphatidylinositol-5-phosphate binding
calcium-release channel activity
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-4-phosphate binding
phosphatidic acid binding
phosphatidylinositol-3,5-bisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bbcf2:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline