This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GKN2 SIRNA Assays ›
Gene Symbol:
GKN2
Gene Name:
gastrokine 2
Gene Aliases:
BRICD1B, GDDR, PRO813, TFIZ1, VLTI465
Chromosome Location:
Chr.2: 68945232 - 68952970 on Build GRCh38
UniGene ID:
Hs.16757
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 216549

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GKN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182536.2 NP_872342.2 3 228
GenBank mRNA AF494509.2 - 3 228
AJ966788.1 - 3 149
AY358664.1 - 3 203
AY943908.1 - 3 151

Back To Top

More Information


Gene Ontology Categories:

Function(s)

regulation of cell proliferation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline