This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FREM2 SIRNA Assays ›
Gene Symbol:
FREM2
Gene Name:
FRAS1 related extracellular matrix protein 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 38687036 - 38887131 on Build GRCh38
UniGene ID:
Hs.253994
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 216587

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FREM2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207361.5 NP_997244.4 24 10039
GenBank mRNA BX538150.1 - 7 1507
BX538304.1 - 20 4003
BX640686.1 - 4 1176
CR933724.1 - 12 2531

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
cell communication
cell adhesion
heart development
inner ear development
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline