siRNA Details
| Gene Symbol: ZNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160183.2 | NP_001153655.1 | 5 | 1961 | ||
| NM_001271637.1 | NP_001258566.1 | 4 | 1828 | |||
| NM_001271638.1 | NP_001258567.1 | 4 | 1835 | |||
| NM_001271639.1 | NP_001258568.1 | 4 | 1824 | |||
| NM_001271640.1 | NP_001258569.1 | 4 | 1883 | |||
| NM_001271649.1 | NP_001258578.1 | 2 | 1619 | |||
| NM_006524.3 | NP_006515.2 | 3 | 1746 | |||
| NR_073391.1 | 5 | 1928 | ||||
| GenBank mRNA | AK027335.1 | - | 1 | 501 | ||
| AK055644.1 | - | 4 | 1788 | |||
| BC141847.1 | - | 4 | 1795 | |||

ZNF138
Genomic Map