This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF839 SIRNA Assays ›
Gene Symbol:
ZNF839
Gene Name:
zinc finger protein 839
Gene Aliases:
C14orf131
Chromosome Location:
Chr.14: 102317377 - 102343174 on Build GRCh38
UniGene ID:
Hs.743359
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 217157

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF839
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267827.1 NP_001254756.1 7 1853
NM_001267828.1 NP_001254757.1 7 1784
NM_018335.4 NP_060805.3 7 1866
XM_006720203.3 XP_006720266.2 7 1750
XM_011536945.1 XP_011535247.1 8 1968
XM_011536946.2 XP_011535248.1 7 1818
XM_011536948.2 XP_011535250.1 7 1594
XM_011536949.2 XP_011535251.1 8 1968
XM_011536950.2 XP_011535252.1 7 1818
XM_017021447.1 XP_016876936.1 6 1716
XM_017021448.1 XP_016876937.1 7 1866
XM_017021449.1 XP_016876938.1 6 1132
XM_017021450.1 XP_016876939.1 8 4049
XM_017021451.1 XP_016876940.1 7 1772
GenBank mRNA AF155112.1 - 5 495
AK001994.1 - 3 282
AK093342.1 - 7 1853
AK125678.1 - 3 1002
AK223312.1 - 3 266
AK289632.1 - 7 1784
AL137478.1 - 6 1140
BC073137.1 - 5 643

Back To Top

More Information


Gene Ontology Categories:

Process(es)

metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline