siRNA Details
| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006962.1 | NP_001006963.1 | 6 | 934 | ||
| XM_006251940.2 | XP_006252002.1 | 6 | 1922 | |||
| XM_006251942.2 | XP_006252004.1 | 6 | 1922 | |||
| XM_006251943.2 | XP_006252005.1 | 6 | 1923 | |||
| XM_006251944.2 | XP_006252006.1 | 6 | 1922 | |||
| GenBank mRNA | BC083766.1 | - | 6 | 934 | 7193196 | |

Tinf2
Genomic Map