This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF484 SIRNA Assays ›
Gene Symbol:
ZNF484
Gene Name:
zinc finger protein 484
Gene Aliases:
BA526D8.4
Chromosome Location:
Chr.9: 92834262 - 92878060 on Build GRCh38
UniGene ID:
Hs.668378
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 217667

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF484
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007101.2 NP_001007102.1 4 1093
NM_001261458.1 NP_001248387.1 4 1163
NM_001261459.1 NP_001248388.1 6 1234
NM_001261460.1 NP_001248389.1 6 1164
NM_031486.2 NP_113674.1 5 1138
XM_006717299.3 XP_006717362.1 5 1159
XM_011519073.2 XP_011517375.1 6 1275
XM_011519074.2 XP_011517376.1 5 1048
XM_011519075.2 XP_011517377.1 6 1204
XM_017015178.1 XP_016870667.1 7 1230
XM_017015179.1 XP_016870668.1 7 1078
XM_017015180.1 XP_016870669.1 5 1008
XM_017015181.1 XP_016870670.1 7 1242
GenBank mRNA AK000772.1 - 1 288
AK091203.1 - 4 1043
AK292997.1 - 7 1222
AK299274.1 - 4 1061
AK309617.1 - 5 1138
AL833202.1 - 5 1108
BC112394.1 - 4 988 40008083
BC112396.1 - 4 988 40008084

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-w8hc8:80/100.66.77.245:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0